Charging Stand for HandyStep® touch, Brand
Catalog # 76338-258
Supplier: BRAND GMBH & CO. MX
Specifications
- Description:Inductive charging stand for HandyStep® touch, without AC adapter
- Cat. no.:76338-258
Specifications
About this item
The charging stand for the HandyStep® touch saves space on the laboratory bench and securely holds the device even with a PD-Tip II attached.
- Can be used for both the HandyStep® touch and the HandyStep® touch S
- Inductive charging technology enables charging without cable clutter
- Simply place the HandyStep® touch into the stand, and the charging process begins
The blue LED status indicator at the base of the charging stand shows the charging status:
LED lit: Device is charging
LED not lit: Device is fully charged
Ordering information: Without AC adapter (needs to be ordered separately).