Density Cube Set of 24
Catalog # 470303-938
Supplier: UNITED SCIENTIFIC SUPPLIES
Specifications
- Description:Density cube set of 24
- Cat. no.:470303-938
Specifications
About this item
Observe the density across different materials and sizes.
- Foam storage space
- Includes a Vernier caliper
This set includes three different sized cubes of eight materials commonly used in density experiments. Each material is represented by a 0.5, 1 and 1.5" cube. Materials included are acrylic, aluminum, brass, copper, nylon, polypropylene, PVC and steel. Set includes a reference table showing density values of individual specimens.