Chamber BRITE Autoclave/sterilizer Cleaner
Catalog # 58619-028
Supplier: Heidolph NA, LLC
Specifications
- Description:Chamber BRITE sterilizer cleaning powder, 10 sachets per box
- Environmentally Preferable:
- Cat. no.:58619-028
- Supplier No.:023212803
Specifications
About this item
Chamber Brite cleaning powder is the easiest way to keep autoclaves clean and free of hard water deposits.
- Simple to use premeasured sachets
- Replaces messy liquid detergents
- Increases sterilization efficiency
- Extends autoclave lifecycle
- Odor free and easy to store
Designed specifically for steam sterilizers, Chamber Brite effectively cleans and descales the reservoir, chamber and tubing in minutes.