L-K Industries Benchmark Heated Oil Centrifuges
Supplier: L-K INDUSTRIES MS
Certificates
About this item
Safety first—Class 1, Division 2 certified models available.
- Programmable digital controls maintain sample temperature and RPM accurately
- Motor controller-integrated timer ensures test method compliance
- Integral mounting flanges allow secure attachment to benchtop, stand, or table
- Conformance: ASTM D91, D96, D893, D1796, D2273, D2709, and D4007
- Shipping weight: 110 lb (50 kg)
These benchtop laboratory centrifuges help you meet the requirements of a variety of ASTM methods for oil testing. Program temperature and speed with digital precision, set motor control-integrated timer for test duration, and start the run. Simple controls save time, automated functions allow you to perform other tasks, and safety features protect the lab.
Choose explosion-proof models for hazardous location operation. Additional safety features include a lid-locking system preventing motor operation when open, emergency power-off palm button, and indicator lamps.
Order sample tubes separately.
Note: All models must be hard-wired by a qualified electrician with 12 AWG wire.
Delivery: Includes four-place rotor with buckets.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX17700-00
- Rotor type:
- Long cone tube type
- Min. speed:
- 200 rpm
- Max. speed:
- 2300 rpm
- Max. RCF:
- 1535 ×g
- Timer range:
- Preset at 10 minutes
- Temperature control:
- Analog with LCD display
- Min. temperature:
- Ambient °C (Ambient °F)
- Max. temperature:
- 100 °C (212 °F)
- Power:
- 120 VAC, 60 Hz
- Tube capacity:
- 100 ml
- Warranty:
- 1 year
Specifications
Frequently Bought Together