Order Entry
Mexico
ContactUsLinkComponent
FMI Series RH Valveless Piston Pump Heads
undefined
FMI Series RH Valveless Piston Pump Heads
  • Inlet connection:
  • 1/4" OD tubing
  • 1/4-28 female
  • 1/4" OD tubing
  • 1/4-28 female
  • 1/4-28 female
  • 1/4" OD tubing
  • 1/4-28 female
  • 1/4" OD tubing
  • Max. pressure:
  • 100 psi
  • 100 psi
  • 100 psi
  • 100 psi
  • 100 psi
  • 100 psi
  • 100 psi
  • 100 psi
  • Min. flow rate:
  • 0.018 ml/min
  • 0.018 ml/min
  • 0.018 ml/min
  • 0.03 ml/min
  • 0.03 ml/min
  • 0.018 ml/min
  • 0.03 ml/min
  • Min. motor RPM:
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • 6 rpm
  • Wetted materials:
  • Ceramic (alumina), ETFE
  • Ceramic (alumina), PVDF
  • Ceramic (alumina), PVDF
  • Ceramic (alumina), ETFE
  • Ceramic (alumina), PVDF
  • Ceramic (alumina), ETFE
  • Ceramic (alumina), ETFE
  • Ceramic (alumina), PVDF
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together