Catalyst Pump Coiled Tubing For FH100, FH100D, FH100X and FH100DX Pumps
Proveedor: Avantor
Certificados
Acerca de este artículo
Four formulations available to cover most chemical compatible applications.
- To help simplify choices, a platinum-cured silicone, two TPEs and a long life PVC tubing material are offered.
- Standard 15 meter lengths
- Available in both thin and thick wall configurations.
- FH100 and FH100D utilize tubing with a wall thickness of 1.6 mm.
- FH100X and FH100DX utilize tubing with a wall thickness of 2.4 mm.
Choose from a wide range of tubing options for your Catalyst pumps.
- Environmentally Preferable:
Frequently Bought Together