Flowline XP88-0 Explosion-Proof Ultrasonic Level Transmitter
Catalog # MFLX68873-81
Supplier: VWR International
Specifications
- Display type:6-digit LCD
- Max. depth:7.5 m (24.6 ft.)
- Max. pressure:2 bar (30 psi)
- Description:Explosion-Proof Ultrasonic Level Transmitter
- Accuracy of depth range:±0.2% of max range
- Dead band:8"
- Loop resistance:500 Ω max at 24 VDC
- Max. process temperature:60 °C
- Min. depth:20 cm (8")
- Min. process temperature:-4 °F
- Resolution of depth range:2 mm (0.079")
- Signal output:4 to 20 mA
- Warranty:1 year
- Cat. no.:MFLX68873-81
- Power:18 - 28 VDC
Specifications
About this item
Explosion-proof ultrasonic level transmitter ideal for hazardous applications.
- Compact XP enclosure with viewing window and dual conduit ports
- Narrow 3'' beam diameter enables flexible installation in applications with limited measurement space
- Rugged corrosion resistant PVDF transducer and process mount for chemical and wastewater
- Fail-safe intelligence with diagnostic feedback for easy troubleshooting
- Display indicates air gap or level height distance in inch or centimeter
EchoSafe™, an explosion proof, two-wire ultrasonic transmitter provides noncontact level measurement up to 10m in hazardous bulk storage, day tank and sump applications, and is well suited for challenging corrosive, slurry or waste media. Media examples include sodium hypochlorite, wastewater and resin. The integral display and push button module enable fast and simple calibration.
- Display type:6-digit LCD
- Max. depth:7.5 m (24.6 ft.)
- Max. pressure:2 bar (30 psi)
- Description:Explosion-Proof Ultrasonic Level Transmitter
- Environmentally Preferable:
- Accuracy of depth range:±0.2% of max range
- Dead band:8"
- Loop resistance:500 Ω max at 24 VDC
- Max. process temperature:60 °C
- Min. depth:20 cm (8")
- Min. process temperature:-4 °F
- Resolution of depth range:2 mm (0.079")
- Signal output:4 to 20 mA
- Warranty:1 year
- Cat. no.:MFLX68873-81
- Power:18 - 28 VDC