Order Entry
Mexico
ContactUsLinkComponent
Stainless Steel Male Threaded Elbow to Push-to-Connect Adapters
Stainless Steel Male Threaded Elbow to Push-to-Connect Adapters
Certificates
undefined
Stainless Steel Male Threaded Elbow to Push-to-Connect Adapters
Specifications
  • Max. operating pressure:
  • 225 psi
  • 225 psi
  • 225 psi
  • Max. temperature:
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • Max. vacuum:
  • 762 mm Hg (30" Hg)
  • 762 mm Hg (30" Hg)
  • 762 mm Hg (30" Hg)
  • Min. temperature:
  • –20 °C (–4 °F)
  • –20 °C (–4 °F)
  • –20 °C (–4 °F)
  • Seal material:
  • FKM
  • FKM
  • FKM
  • Sterilisation:
  • Not recommended
  • Not recommended
  • Not recommended
  • Warranty:
  • 1 year
  • 1 year
  • 1 year