Order Entry
Mexico
Orders LinkContactUsLinkComponent
Suction Line Pump Filter Strainers
Supplier:   SPECIALTY MFG CO
Suction Line Pump Filter Strainers
Supplier:  SPECIALTY MFG CO
Certificates
Suction Line Pump Filter Strainers
19 Options Available Below
Product Details & Documents
Protect pump from sediments filtering into suction line.
  • PVC (polyvinyl chloride) or Nylon base material of construction
  • 20, 40, 80 mesh sizes available
  • Easily removable for cleaning or replacing
These suction line pump filter strainers are ideal for keeping sediment and debris out of your pump for optimal performance. Choose from PVC (polyvinyl chloride) or nylon base material in a wide range of mesh sizes for any application.
Specifications for Product Family
Specifications
  • Outlet connection:
  • 0.25 NPT(F)
  • 0.25 NPT(F)
  • 0.375 NPT(F)
  • 0.375 NPT(F)
  • 0.375 NPT(F)
  • 0.5 NPT(F)
  • 0.5 NPT(F)
  • 0.5 NPT(F)
  • 0.5 NPT(F)
  • 0.75 NPT(F)
  • 0.75 NPT(F)
  • 0.75 NPT(F)
  • 1 NPT(F)
  • 1 NPT(F)
  • 1 NPT(F)
  • 1.5 NPT(F)
  • 1.5 NPT(F)
  • 1.5 NPT(F)
  • 0.75 NPT(F)
  • 0.75 NPT(F)
  • Wetted materials:
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • PVC and Stainless Steel
  • Nylon and Stainless Steel
  • PVC and Stainless Steel
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 6 Months
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 6 Months
  • 1 year
  • 1 year
  • 6 Months
  • 1 year
  • 1 year
  • 6 Months
Specifications
Product Family Options
Product Information
  • Inlet Connection
    Availability
    Price
  • MFLX74604-02
    Suction Line Strainer, PVC, 1/4" NPT(F), 40 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1/4" NPT(F), 40 Mesh (stainless steel)
    0.25 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $213.00
    $213.00
    Each (1UN)
       $213.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-02
    Outlet connection
    0.25 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-04
    Suction Line Strainer, PVC, 1/4" NPT(F), 80 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1/4" NPT(F), 80 Mesh (stainless steel)
    0.25 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $213.00
    $213.00
    Each (1UN)
       $213.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-04
    Outlet connection
    0.25 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-10
    Suction Line Strainer, PVC, 3/8" NPT(F), 20 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/8" NPT(F), 20 Mesh (stainless steel)
    0.375 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $221.00
    $221.00
    Each (1UN)
       $221.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-10
    Outlet connection
    0.375 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-12
    Suction Line Strainer, PVC, 3/8" NPT(F), 40 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/8" NPT(F), 40 Mesh (stainless steel)
    0.375 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $221.00
    $221.00
    Each (1UN)
       $221.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-12
    Outlet connection
    0.375 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-14
    Suction Line Strainer, PVC, 3/8" NPT(F), 80 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/8" NPT(F), 80 Mesh (stainless steel)
    0.375 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $221.00
    $221.00
    Each (1UN)
       $221.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-14
    Outlet connection
    0.375 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-20
    Suction Line Strainer, PVC, 1/2" NPT(F), 20 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1/2" NPT(F), 20 Mesh (stainless steel)
    0.5 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $246.00
    $246.00
    Each (1UN)
       $246.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-20
    Outlet connection
    0.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-22
    Suction Line Strainer, PVC, 1/2" NPT(F), 40 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1/2" NPT(F), 40 Mesh (stainless steel)
    0.5 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $246.00
    $246.00
    Each (1UN)
       $246.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-22
    Outlet connection
    0.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-23
    Suction Line Strainer, PVC, 1/2" NPT(F), 50 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1/2" NPT(F), 50 Mesh (stainless steel)
    0.5 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $246.00
    $246.00
    Each (1UN)
       $246.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-23
    Outlet connection
    0.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    6 Months
  • MFLX74604-24
    Suction Line Strainer, PVC, 1/2" NPT(F), 80 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1/2" NPT(F), 80 Mesh (stainless steel)
    0.5 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $246.00
    $246.00
    Each (1UN)
       $246.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-24
    Outlet connection
    0.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-30
    Suction Line Strainer, PVC, 3/4" NPT(F), 20 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/4" NPT(F), 20 Mesh (stainless steel)
    0.75 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $281.00
    $281.00
    Each (1UN)
       $281.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-30
    Outlet connection
    0.75 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-32
    Suction Line Strainer, PVC, 3/4" NPT(F), 40 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/4" NPT(F), 40 Mesh (stainless steel)
    0.75 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $281.00
    $281.00
    Each (1UN)
       $281.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-32
    Outlet connection
    0.75 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-34
    Suction Line Strainer, PVC, 3/4" NPT(F), 80 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/4" NPT(F), 80 Mesh (stainless steel)
    0.75 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $293.00
    $293.00
    Each (1UN)
       $293.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-34
    Outlet connection
    0.75 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-42
    Suction Line Strainer, PVC, 1" NPT(F), 40 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1" NPT(F), 40 Mesh (stainless steel)
    1 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $437.00
    $437.00
    Each (1UN)
       $437.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-42
    Outlet connection
    1 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-43
    Suction Line Strainer, PVC, 1" NPT(F), 50 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1" NPT(F), 50 Mesh (stainless steel)
    1 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $437.00
    $437.00
    Each (1UN)
       $437.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-43
    Outlet connection
    1 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    6 Months
  • MFLX74604-44
    Suction Line Strainer, PVC, 1" NPT(F), 80 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1" NPT(F), 80 Mesh (stainless steel)
    1 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $437.00
    $437.00
    Each (1UN)
       $437.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-44
    Outlet connection
    1 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-52
    Suction Line Strainer, PVC, 1-1/2" NPT(F), 40 Mesh stainless steel
    Suction Line Strainer, PVC, 1-1/2" NPT(F), 40 Mesh stainless steel
    1.5 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $504.00
    $504.00
    Each (1UN)
       $504.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-52
    Outlet connection
    1.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74604-53
    Suction Line Strainer, PVC, 1-1/2" NPT(F), 50 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1-1/2" NPT(F), 50 Mesh (stainless steel)
    1.5 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $533.00
    $533.00
    Each (1UN)
       $533.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-53
    Outlet connection
    1.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    6 Months
  • MFLX74604-54
    Suction Line Strainer, PVC, 1-1/2" NPT(F), 80 Mesh (stainless steel)
    Suction Line Strainer, PVC, 1-1/2" NPT(F), 80 Mesh (stainless steel)
    1.5 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $533.00
    $533.00
    Each (1UN)
       $533.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-54
    Outlet connection
    1.5 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    1 year
  • MFLX74605-12
    Suction Line Strainer, Nylon, 3/4" NPT(F), 40 Mesh (stainless steel)
    Suction Line Strainer, Nylon, 3/4" NPT(F), 40 Mesh (stainless steel)
    0.75 NPT(F)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $701.00
    $701.00
    Each (1UN)
       $701.00
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74605-12
    Outlet connection
    0.75 NPT(F)
    Wetted materials
    Nylon and Stainless Steel
    Warranty
    1 year
  • MFLX74604-33
    Suction Line Strainer, PVC, 3/4" NPT(F), 50 Mesh (stainless steel)
    Suction Line Strainer, PVC, 3/4" NPT(F), 50 Mesh (stainless steel)
    0.75 NPT(F)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX74604-33
    Outlet connection
    0.75 NPT(F)
    Wetted materials
    PVC and Stainless Steel
    Warranty
    6 Months