Order Entry
Mexico
ContactUsLinkComponent
CPC® Metal Quick-Disconnect Fittings, Panel-Mount Hose Barb Bodies
undefined
CPC® Metal Quick-Disconnect Fittings, Panel-Mount Hose Barb Bodies
  • Length:
  • 5.08 cm
  • 5.08 cm
  • 5.08 cm
  • 5.08 cm
  • Max. operating pressure:
  • 250 psi
  • 250 psi
  • 250 psi
  • 250 psi
  • 250 psi
  • Max. temperature:
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • Min. temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • Seal material:
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Valve spring material:
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • Warranty:
  • 3 Months
  • 3 Months
  • 1 year
  • 3 Months
  • 3 Months
Frequently Bought Together