Order Entry
Mexico
ContactUsLinkComponent
Flux® Drum Pump Tube Sets, PVDF
Flux® Drum Pump Tube Sets, PVDF
Supplier:  FLUX PUMPS CORP
Certificates
undefined
Flux® Drum Pump Tube Sets, PVDF
Supplier:  FLUX PUMPS CORP
Specifications
  • Max. temperature:
  • 212 °C
  • 212 °C
  • Outlet connection:
  • 3/4" hose barb
  • 3/4" hose barb
  • Tube ext.Ø, inlet:
  • 4 cm (1.5625")
  • 4 cm (1.5625")
  • Tube length, inlet:
  • 47"
  • 39"
  • Wetted materials:
  • PVDF body; Tefzel impeller, Viton, PTFE, carbon, Hastelloy C
  • PVDF body; Tefzel impeller, Viton, PTFE, carbon, Hastelloy C
  • Warranty:
  • 6 Months
  • 6 Months