Order Entry
Mexico
Orders LinkContactUsLinkComponent
Thiomersal 97.0-101.0%

Thiomersal 97.0-101.0%

Glass bottle for solids, 100 g
Catalog # TS11868-1000
Toxic
Health
Environment
Supplier #:  11868-1000
Thiomersal 97.0-101.0%
Catalog # TS11868-1000
CAS Number:  54-64-8
Thiomersal 97.0-101.0%
Thiomersal 97.0-101.0%

Glass bottle for solids, 100 g

CAS Number:  54-64-8
Price
$14,120.00
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Ethylmercurithiosalicylic acid sodium salt, Sodium ethylmercurithiosalicylate, Thimerosal, Merthiolat, 2-[(Ethylmercury)thio]benzoic acid sodium salt, 2-(Ethylmercuriomercapto)benzoic acid sodium salt, 2-(Ethylmercuriothio)benzoic acid sodium salt
  • CAS Number:54-64-8
  • Formula:2-(C₂H₅HgS)C₆H₄CO₂Na
  • MDL Number:MFCD00013062
  • Molecular Weight:404.82 g/mol
  • Merck Index:12,09451
  • Melting Pt:232…233 °C (dec.)
  • Flash Pt:250 °C
  • Storage Temperature:Ambient
  • EINECS:200-210-4
  • UN:2025
  • ADR:6.1,II
Specifications

Specification Test Results

  • Appearance:
    White to cream Powder
  • Infrared spectrum:
    Conforms
  • Assay:
    97.0 to 101.0 % (On dried substance)
  • Loss on drying:
    ≤0.5 % (vacuum/P2O5)
  • Additional info:
    ion mercuric: ≤ 0.7%
  • pH:
    6 to 8 (1 % solution)

Specifications

  • Pack type:
    Glass bottle for solids
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.