Masterflex® Dispensing Tips, Polypropylene, Avantor®
Polypropylene dispensing tip with Luer lock for ¹⁄₁₆" and ⅛" ID tubing., Female Luer to Barb, Straight, PP
MFLX78226-62
: 78226-62
Attach with just a twist!
- Constructed of durable polypropylene
- Easily attach flexible tubing to the hose barb connection
These luer fittings are ideal for a variety of critical applications, including connection to syringes or small diameter flexible tubing.
- Connection Type:Female Luer to Barb
- Flow pattern:Straight
- Description:Polypropylene dispensing tip with Luer lock for ¹⁄₁₆" and ⅛" ID tubing.
- Fitting Material:PP