Masterflex® I/P® Variable-Speed Air-Powered Pump Systems, Avantor®
I/P ATEX Zone 2 Air-Powered Pump System, 100-650 rpm., 8000 ml/min, 228.6 mm (9"), 333.5 mm (13.13"), 215.9 mm (8.5"), 77602-80 (I/P® Easy-Load® ATEX head), 650 rpm, Painted steel
Catalog # MFLX77980-10
CE Compliant
Supplier: Avantor Fluid Handling
Supplier #: 77980-10
Masterflex® I/P® Variable-Speed Air-Powered Pump Systems, Avantor®
Catalog # MFLX77980-10
Supplier: Avantor Fluid Handling
CE Compliant
Jump to:
Product Details & Documents
Pump safely in hazardous
environments.
- Flow rate 0.6 to 13 LPM I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
- Easily adjust motor speed by using included regulator
- Effortlessly connect to your existing air supply or compressor
- The 3/4-hp air-powered motor starts smooth, runs cool, and requires little maintenance
- ATEX Zone 2 approved; rated to EEXII 3 G c IIC T6; NEC Class 1, Division 2, Groups A, B, C, D, T6
Use this variable-speed air-powered drive when electrical power may be unsafe or impractical. For safe operation, carefully ground to protect from static electricity.
Specifications
- Maximum Flow Rate:8000 ml/min
- Description:I/P ATEX Zone 2 Air-Powered Pump System, 100-650 rpm.
- Height:228.6 mm (9")
- Length:333.5 mm (13.13")
- Max. RPM:650 rpm
- Pump Head:77602-80 (I/P® Easy-Load® ATEX head)
- Width:215.9 mm (8.5")
- Housing Material:Painted steel
- Min. flow rate:1200 ml/min
- Open-head sensor:No
- Warranty:2 years
- No. of channels:1
- Control type:Variable-Speed Air-Powered
- No. of rollers:3
- Cat. No.:MFLX77980-10
- Accuracy:±10%
- Drive IP rating:IP 34
- Tubing:06434-73, 10-ft (C-FLEX® Ultra, I/P® 73)
- Duty cycle:Continuous
- Pump series:I/P
- Model:77980-10
- Drive:07589-30 (I/P® Variable-Speed Air-Powered Drive)
- Tubing sizes accepted:I/P® 26, 73, 82
- Washdown capable:No
- Min. RPM:100 rpm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



