Ammonium peroxodisulphate (APS) 150 mg, tablets, Biotechnology Grade
97064-852
: VWR
:
- Size:100 tablets
- Environmentally Preferable:
- Cat. no.:97064-852
- Supplier No.:K833-100TABS
- Dissolution Time (1 tablet/1mL Water):<= 4 minutes
- Polymerization Time:<= 30 minutes
- Solubility (10%, Water):PASS
- Tablet Weight:112 - 137 mg
: Peroxidisulphuric acid diammonium salt, Ammonium persulphate
:
:228.2 g/mol
:Ambient
:7727-54-0
:(NH₄)₂S₂O₈
:MFCD00003390
:120
:13,00545