Order Entry
Mexico
ContactUsLinkComponent
Vikan® Single-Blade Ultra-Hygiene Squeegees, Remco
  76530-374
undefined
Vikan® Single-Blade Ultra-Hygiene Squeegees, Remco
  76530-374
 :  71505
  • Color:
    White
  • Cat. no.:
    76530-374
  • Blade Width:
    20"
The one-piece, single-blade squeegee has an ultra-hygienic design that minimizes areas for water and food soils to collect. This seamless, one-piece construction is especially effective in areas requiring the highest level of hygiene.
  • Rubber blade, polypropylene block
  • Seamless design
  • Euro (hygienic) thread type
  • Available in three blade widths
The soft and flexible rubber blade is ideal for removing water or food debris from floors and food contact surfaces. Hygiene squeegees are a good foundation for any HACCP or food safety program.