Order Entry
Mexico
Orders LinkContactUsLinkComponent
DNA Model - Preassembled
DNA model
  470224-018
 :  DNAM01
DNA Model - Preassembled
  470224-018
DNA Model - Preassembled
DNA Model - Preassembled
DNA model
$2,422.53
Each (1UN)
Built to scale.
  • Easy to connect pieces
  • Shows 16 base pairs
This pre-assembled DNA model is built to a scale of 10 cm to 1 nm and accurately depicts a 16 base pair section of DNA. The phosphatedeoxyribose backbone and the four bases are represented by custom plastic moldings that show the shapes of the molecules in scale and illustrate the bonding between them. The model is supported by a central pillar which can rotate on a circular stand.
Dimensions of the model are 24H×8" Ø, making it suitable for classroom demonstrations.
 : Small parts: Not for children under three years.
  • Description:
    DNA model