Order Entry
Mexico
Orders LinkContactUsLinkComponent
BioX Serological Pipettes
BioX Serological Pipettes
Serological pipette, 5 ml, Individually wrapped
Catalog # 76469-342
Supplier #:  BX07-5005
BioX Serological Pipettes
Catalog # 76469-342
BioX Serological Pipettes
BioX Serological Pipettes
Serological pipette, 5 ml, Individually wrapped
Price
$1,612.60
Log in for availability
Case (200UN)
Product Details & Documents
These high quality serological pipettes are DNase/RNase free and come individually wrapped.
  • Sterile
  • Individually wrapped, paper/plastic wrap
  • Non-pyrogenic
  • DNase/RNase free
  • Color coded
These sterile serological pipettes are not only DNase/RNase free and non pyrogenic, but they are also accurate to ±2% and color coded for your convenience.
Specifications
  • Capacity:
    5 ml
  • Description:
    Serological pipette
  • Packed:
    Individually wrapped