Model 2500 Water Bath Dissolution System, Distek
: Avantor
The model 2500 dissolution system offers great flexibility and configurability. It can be configured as USP Apparatus 1, 2, 5 and 6 plus intrinsic dissolution. The extensive users, method and reporting storage capabilities are enabled by a brilliant touch screen.
- Volume flexibility
- Self-priming circulator/heater
- Electronic qualification guidance
- Color touch screen display
- Quick change paddles and baskets
- Automatic height adjustment
- Wake up and sleep mode
Unparalleled control of the instrument plus automatic storage of component serial numbers and automatic qualification reminders make the Model 2500 robust and reliable enough for QC applications yet flexible enough for R&D use.
: ETL, CE.
: Does not include installation or qualification services.
: Includes shafts, vessels, paddles, baskets, basket adapters, vessel covers, and a one-year comprehensive warranty.
Frequently Bought Together