Heat Pack
10 Hour hand warmer, 5.1×10.2 cm (2×4″)
Catalog # 470127-432
Supplier: 3G PACKAGING, INC.
Supplier #: 10 HOUR
Jump to:
Product Details & Documents
Disposable hand warmers - ideal for outdoor workers and those who engage in cold weather sporting activities.
- Effective for arthritic or muscle/joint pain
- Soft, lightweight and easy to activate
- Maintains temperatures of up to 65° Celsius (150° Fahrenheit) for up to 10 hours
- Easily fits in pocket, purse, knapsack or glove compartment
Specifications
- Description:10 Hour hand warmer
- Dimensions:5.1×10.2 cm (2×4″)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



