Avantor® ACE® C4-300, HPLC Columns, 5 µm
76390-704
: Avantor
- Column Format:Analytical
- Ø int.:4.60 mm
- Particle Size:5 µm
- Pore size:300 Å
- Max. pressure:275 bar
- Description:¹/₁₆" connections
- Length:150 mm
- Endcapped:Yes
- USP code:L26
- pH range:2 - 8
- Surface area:100 m²/g
- Carbon load:2.60%
- Cat. no.:76390-704
- Phase:C4
Avantor® ACE® C4-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
- Designed for peptide and protein separations
- Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.