BioClean™ Advance BASL Sterile Latex Cleanroom Gloves, Ansell
Sterile latex cleanroom gloves.
- Flexible and comfortable
- Easy double-donning
BioClean™ advance gloves are anatomically shaped providing flexibility and wearer comfort. The natural-colored long gloves have been designed to be double-donnable, feature a textured surface for enhanced grip and a beaded cuff for stability on the arm.
: EN 420:2003 + A1:2009; EN 420:2003 + A1:2009; EN 455 Part 2; ISO 9001:2015 and gloves are compatible with Class 10 (ISO 4) environments.
: One pair per inner PE wallet; one wallet per sealed Non-particulating EasyTear™ EasyTear PE pouch; 10 pouches per sealed outer PE bag; 20 outer bags per lined carton (200 pairs).
- Catalog No:
- 75982-318
- 75982-320
- 75982-324
- 75982-348
- 75982-326
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured
- Textured
- Textured
- Textured
- Textured
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
Frequently Bought Together