Order Entry
Mexico
ContactUsLinkComponent
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, 2 µm
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, 2 µm
  76392-574
 :  Avantor
undefined
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, 2 µm
  76392-574
 :  Avantor
 :  EXL-1011-1503U

 

  • Column Format:
    Analytical
  • Ø int.:
    3.00 mm
  • Particle Size:
    2 µm
  • Pore size:
    90 Å
  • Max. pressure:
    1000 bar (14504 psi)
  • Length:
    150 mm
  • Endcapped:
    Yes
  • USP code:
    L1
  • pH range:
    1.5 - 11.5
  • Surface area:
    400 m²/g
  • Carbon load:
    14.8%
  • Cat. no.:
    76392-574
  • Phase:
    SuperC18™

 

 

Avantor® ACE® Excel® SuperC18™ columns provide high stability across an extended pH range. These columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.

  • Ultra-inert for maximum performance and reproducibility
  • Ultra low bleed for improved LC/MS compatibility
  • Wide range of particle sizes (1.7 to 10 µm) and complementary phases

These columns are made of stainless steel.