Order Entry
Mexico
ContactUsLinkComponent
Avantor® ACE® C18 Ultra-Inert, Analytical HPLC Columns, 3 µm
Avantor® ACE® C18 Ultra-Inert, Analytical HPLC Columns, 3 µm
  76382-588
 :  Avantor
   Low Manufacturing Impact
undefined
Avantor® ACE® C18 Ultra-Inert, Analytical HPLC Columns, 3 µm
  76382-588
 :  Avantor
 :  ACE-211-1546
   Low Manufacturing Impact

 

  • Column Format:
    Analytical
  • Ø int.:
    4.60 mm
  • Particle Size:
    3 µm
  • Pore size:
    300 Å
  • Max. pressure:
    400 bar (5802 psi)
  • Description:
    ¹/₁₆" connections
  • Length:
    150 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001:2015 (530840 UM15)
  • Endcapped:
    Yes
  • USP code:
    L1
  • pH range:
    2 - 8
  • Surface area:
    100 m²/g
  • Carbon load:
    9.0%
  • Cat. no.:
    76382-588
  • Phase:
    C18

 

 

Avantor® ACE® C18-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.

  • Designed for peptide and protein separations
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3 to 10 µm) and complementary phases

These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules. These columns provide excellent peak shape even at extremely low levels of additives such as TFA, making them ideal for LC/MS applications.