Order Entry
Mexico
ContactUsLinkComponent
Kimble® KEM-KIT® Williamson Macroscale Standard Taper Kit, DWK Life Sciences
Kimble® KEM-KIT® Williamson Macroscale Standard Taper Kit, DWK Life Sciences
Catalog # KT269330-0000
undefined
Kimble® KEM-KIT® Williamson Macroscale Standard Taper Kit, DWK Life Sciences
Catalog # KT269330-0000
Supplier Number:  269330-0000
Specifications
  • Description:
    KIMBLE® KEM-KITS® Williamson macroscale laboratory glass kit, [ST] 14/20
  • Cat. no.:
    KT269330-0000
  • Supplier No.:
    269330-0000
Specifications
About this item
This KEM-KIT® designed to address the experiments detailed in the Williamson text, macroscale and microscale organic experiments, published by D.C. Heath and Company, Lexington, Massachusetts.
  • Smaller quantities afford a greater degree of safety, improve air quality in the lab and substantially reduce disposal costs
  • Separatory funnel for extractions and for liquid addition in reaction assemblies
  • Classical atmospheric, steam, fractional and vacuum distillations are possible with kit components
  • Apparatus set-up time is minimal, providing the student with more lab time
Certifications: Glassware manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.