Order Entry
Mexico
ContactUsLinkComponent
GelStar™ Nucleic Acid Gel Stain 10,000X, Lonza
GelStar™ Nucleic Acid Gel Stain 10,000X, Lonza
Catalog # 12001-804
CAS Number:  
undefined
GelStar™ Nucleic Acid Gel Stain 10,000X, Lonza
Catalog # 12001-804
Supplier Number:  50535
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    GelStar™ Nucleic Acid Gel Stain 10000X
  • Size:
    2×250 μl
  • Cat. no.:
    12001-804
  • Supplier No.:
    50535
Specifications
About this item
GelStar is a sensitive fluorescent stain for detecting both DNA and RNA.
  • Maximum sensitivity - detect as little as 20 pg of dsDNA or 3 ng of RNA,
  • Versatile - use for agarose or polyacrylamide gel electrophoresis,
  • Ideal alternative to silver staining
GelStar™ Nucleic Acid Gel Stain is a highly sensitive fluorescent stain for detecting both DNA and RNA. You can choose to either add GelStarTM Stain to your agarose solution prior to casting or to post stain your gels. GelStarTM Nucleic Acid Gel Stain exhibits exceptional signal-to-noise ratio with minimal background. Maximum sensitivity, detect as little as 20 pg of dsDNA or 3 ng of RNA