Order Entry
Mexico
ContactUsLinkComponent
Swift M3600 Series Compound Microscopes
Swift M3600 Series Compound Microscopes
Catalog # 470317-656
Supplier:  Swift Instruments
undefined
Swift M3600 Series Compound Microscopes
Catalog # 470317-656
Supplier:  Swift Instruments
Supplier Number:  M3604C-4
Specifications
  • Model:
    M3604C-4
  • Description:
    4×, 10×, 400×, 1000× with 1.25 Abbe Condenser and Built-In Mech. Stage
  • Cat. no.:
    470317-656
  • Illumination:
    Cordless LED
Specifications
About this item
This microscope series is perfect for high school students.
  • Lead-free objectives
  • Coaxial coarse and fine focus
  • Variable LED illumination
  • Dimmer switch
  • Built-in camera port
Now with coaxial focusing and upgraded LED illumination, the M3600 Series continues the Swift tradition for innovation, quality, and student-proof features.