Adson-Brown Tissue Forceps
Adson Brown Forceps, Stainless steel, 115 mm (4.5")
Catalog # 470315-180
Supplier: DR INSTRUMENTS INC. SE
Supplier #: 13AB
Jump to:
Product Details & Documents
Easily manipulate delicate tissue.
- Broad control surface
- High quality stainless steel
Using narrow tips, and a broad thumb control surface, Adson-Brown forceps give you excellent control when handling delicate tissue. The tips are designed to minimize damage to tissue applying pressure evenly over the full contact surface.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Description:Adson Brown Forceps
- Length:115 mm (4.5")
- Material:Stainless steel
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



