Order Entry
Mexico
ContactUsLinkComponent
Trisodium phosphate dodecahydrate ≥98% ACS, Fluka™
Trisodium phosphate dodecahydrate ≥98% ACS, Fluka™
Catalog # BJ222003-500G
CAS Number:  
Trisodium phosphate dodecahydrate ≥98% ACS, Fluka™
Catalog # BJ222003-500G
Supplier Number:  222003-500G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 g
  • Cat. no.:
    BJ222003-500G
  • Supplier No.:
    222003-500G

Specifications

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • Insoluble matter:
    Max. 0.01 %
  • Free alkali (as NaOH):
    Max. 2.5 %
  • Iron (Fe):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.01 %

Specification Test Results

About this item

 :
 :380.12 g/mol
Melting Pt:75 °C (dec.)
 :GHS07
Storage Temperature:Ambient
CAS Number:10101-89-0
Formula:Na₃PO₄·12H₂O
MDL Number:MFCD00149198