Molybdic acid ≥85% (MoO₃ basis) ACS, Fluka™
Catalog # BJ232084-100G
Supplier: HONEYWELL INTERNATIONAL INC MX
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:100 g
- Cat. no.:BJ232084-100G
- Supplier No.:232084-100G
Specifications
Specification Test Results
- Assay:Min. 85.0 %
- Insoluble in Ammonia Solution:Max. 0.01 %
- Heavy metals (as Pb):Max. 0.003 %
- Phosphate, Arsenate, Silicate (as SiO₂):Max. 0.001 %
- Chloride (Cl):Max. 0.002 %
- Phosphate (PO₄):Max. 5 ppm
- Sulfate (SO₄):Max. 0.2 %
Specification Test Results
About this item
:
:161.95 g/mol
Storage Temperature:Ambient
EINECS:231-970-5
CAS Number:7782-91-4
Formula:H₂MoO₄
MDL Number:MFCD00036287