Order Entry
Mexico
Orders LinkContactUsLinkComponent
Buchner Filtering Flasks
Buchner Filtering Flasks
Buchner Filtering Flasks
Certificates
Buchner Filtering Flasks
Buchner Filtering Flasks
8 Options Available Below
Product Details & Documents
Buchner filtering flasks are made of high quality, heavy duty borosilicate glass.
Ordering information: Stopper not included.
Product Family Options
Product Information
  • Capacity
    Diameter ×H
    Availability
    Price
  • 470313-976
    Socket Size 19/26, Side Socket Arm 0.75"
    Socket Size 19/26, Side Socket Arm 0.75"
    100 ml
    2.5×4"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $369.25
    $369.25
    Each (1UN)
       $369.25
    Supplier Number: CH0422A
  • 470313-978
    Socket Size 24/29, Side Socket Arm 0.75"
    Socket Size 24/29, Side Socket Arm 0.75"
    100 ml
    2.5×4"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $459.52
    $459.52
    Each (1UN)
       $459.52
    Supplier Number: CH0422B
  • 470313-980
    Socket Size 24/29, Side Socket Arm 0.90"
    Socket Size 24/29, Side Socket Arm 0.90"
    250 ml
    3.5×5"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $543.37
    $543.37
    Each (1UN)
       $543.37
    Supplier Number: CH0422C
  • 470313-982
    Socket Size 29/32, Side Socket Arm 0.90"
    Socket Size 29/32, Side Socket Arm 0.90"
    250 ml
    3.5×5"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $412.24
    $412.24
    Each (1UN)
       $412.24
    Supplier Number: CH0422C01
  • 470313-984
    Socket Size 24/29, Side Socket Arm 1"
    Socket Size 24/29, Side Socket Arm 1"
    500 ml
    4.25×6.5"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $480.47
    $480.47
    Each (1UN)
       $480.47
    Supplier Number: CH0422D
  • 470313-986
    Socket Size 29/32, Side Socket Arm 1"
    Socket Size 29/32, Side Socket Arm 1"
    500 ml
    4.25×6.5"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $604.08
    $604.08
    Each (1UN)
       $604.08
    Supplier Number: CH0422D01
  • 470313-988
    Socket Size 24/29,
    Socket Size 24/29,
    1000 ml
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,028.70
    $1,028.70
    Each (1UN)
       $1,028.70
    Supplier Number: CH0422E
  • 470313-990
    Socket Size 29/32
    Socket Size 29/32
    1000 ml
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $17.80
    $17.80
    Clearance
    Each (1UN)
       $17.80
    Supplier Number: CH0422F