Glass Wool
Glass Wool Pack of 500 g
Catalog # 470313-974
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: GLW500
Jump to:
Product Details & Documents
For filtering out fine precipitates when filter papers are unsuitable.
- Non-flammable
- Provides thermal insulation
Non-flammable glass wool for use in general laboratory and industrial applications. Used for filtering out fine precipitates where filter papers are not suitable and the intertwined borosilicate glass fibers provide thermal insulation.
Specifications
- Description:Glass Wool Pack of 500 g