Alcohol Burner Dual Wick
Catálogo # 470313-338
Supplier: GSC INTERNATIONAL SE
Specifications
- Description:Alcohol burner with dual wick
- Cat. no.:470313-338
Specifications
About this item
This all metal, permanent wick alcohol burner has a tank capacity of 100 ml.
The 'wick' portion is a ceramic fiber inserted into a thin copper tube. This construction allows for a constant flame. The burner is fueled by high-quality alcohol which can be found in most local hardware stores (not drug store rubbing alcohol). This is an excellent alternative to the traditional glass alcohol burners because it is durable and less likely to spill. Instructions included.