$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
Tapes
Tape, KD11, Caplugs
Tape, KD11, Caplugs
76247-866
:
Caplugs
Tape, KD11, Caplugs
76247-866
:
Caplugs
:
D55158-01
Description:
Tape KD11, 18" Liner
Dimensions:
17.5×2160"
Cat. no.:
76247-866