Zinc Plated Steel Handle Applicator Brush, Gordon Brush
76243-700
- Bristle Material:Horse hair
- Handle Material:Steel, zinc plated
- Description:Zinc Plated Steel Brush
- Brush Head Width:5/8"
- Cat. no.:76243-700
- Handle Length:41/2"
- Trim Length:¾"
This zinc plated steel handle applicator brush is an inexpensive utility duster/cleaner that is an excellent replacement for disposable acid brushes that quickly shed their fibers.
- Anti-static
- Adhesive-free to prevent leaching and contamination of surface areas
- Brush fibers are secured using a zinc plated steel handle, substituting adhesives with 5 tons of mechanical clamping