SRK® Collection and Transport System 922C
Catalog # 35001-166
Supplier: COPAN DIAGNOSTICS INC
Specifications
- Sterility:Sterile
- Description:10 cm traditional polyester swab with skirted tube (16×100 mm) containing 10 ml COPAN SRK® solution
- Cat. no.:35001-166
Specifications
About this item
COPAN’s SRK® system combines a swab with a rinse solution or broth in a convenient plastic tube.
- 10 cm traditional polyester swab with skirted tube containing 10 ml COPAN SRK® solution
- With plastic green capture cap
- Individually wrapped, sterile
The environmental sampling line helps food technologists and quality control managers assess microbial presence on surfaces and equipment.
Delivery: 500 pcs per carton (10 boxes of 50 pcs).