Order Entry
Mexico
ContactUsLinkComponent
Sulzer Mixpac QUADRO™ Mixer with Bayonet, Ellsworth Adhesives
Sulzer Mixpac QUADRO™ Mixer with Bayonet, Ellsworth Adhesives
Catalog # 76240-674
Sulzer Mixpac QUADRO™ Mixer with Bayonet, Ellsworth Adhesives
Catalog # 76240-674
Supplier Number:  EAMAQ05-24LS
Specifications
  • Description:
    Sulzer Mixpac QUADRO™ Mixer with Bayonet
  • Cat. no.:
    76240-674
  • Diameter ×L:
    5.3×115 mm
Specifications
About this item
Sulzer Mixpac QUADRO™ Mixer with Bayonet is a disposable mixer for 25 to 75 ml bayonet style cartridges that can be used with all types of resins such as adhesives, sealing materials, coatings, and foams.
Stepped tips can be cut to make a larger outlet. Outlet diameters are 1.8, 2.8, 3.2, 3.6 and 3.9 mm.