Shoe Cleaner, Ultra-Clean Products
Catalog # 76268-110
Supplier: ULTRA-CLEAN PRODUCTS
Specifications
- Description:Automatic Shoe Cleaner, 1600SA
- Cat. no.:76268-110
Specifications
About this item
Fully automatic shoe cleaner for use with external vacuum systems.
- Simple to operate
- Rugged design gives long life
- Large footprint for stability
- Speed control, brush motor
Five brushes for complete contamination removal.
Vacuum Exhaust - 1.50 I.D. x 1.75 O.D.
Size of main unit - 18"W x 27"L x 10.5"H
Overall height with handle - 40"
Weight - 50 LBS
Shipping Weight - 58 LBS
Voltage - 110-120V AC