Order Entry
Mexico
ContactUsLinkComponent
Inserts for Target DP™ 9-425 Glass Screw-Thread Vials, Thermo Scientific
  66030-902
 :  Avantor
Inserts for Target DP™ 9-425 Glass Screw-Thread Vials, Thermo Scientific
  66030-902
 :  Avantor
  • Cap included:
    No
  • Capacity:
    0.40 ml
  • Description:
    MicroSert™ insert, fill marking
  • Material:
    Glass
  • Cat. no.:
    66030-902
  • Bottom Style:
    Flat
  • Supplier No.:
    C4011-631
The polyspring microseal inserts self-align and provide a cushion against needle contact.
Precision point inserts minimize residual sample loss. Pulled point inserts provide an economical choice for noncritical applications.