Order Entry
Mexico
Orders LinkContactUsLinkComponent
Water Sampler

Water Sampler

Water Sampler, 1.5 L
Catalog # 470308-880
Supplier #:  15010
Water Sampler
Catalog # 470308-880
Water Sampler
Water Sampler

Water Sampler, 1.5 L

Price
$2,770.58
Log in for availability
Each (1UN)
Product Details & Documents

Water sampler for samples of lake, river, or stream water.

  • Collect water samples for quality testing

Efficient, economical water sampler. A single line deploys and triggers the sampler from any depth in lakes, rivers, or ponds. The flow-through design allows a valid sampling for measuring temperature and water quality.

Delivery information: A 14 m line is included.

Caution: Volume may vary slightly. It is recommended that users verify the exact volume of the bottle prior to use.

Specifications
  • Description:
    Water Sampler
  • Nominal Capacity:
    1.5 L