Radiation Can Kit
Radiation can kit
Catalog # 470304-034
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: RDCNKIT
Jump to:
Product Details & Documents
High Quality for the Classroom.
- Includes activity guide
Demonstrate how two identically sized cans absorb and radiate energy differently. One of the screw cap cans is smooth metal and the other is painted dull black.
Water cools or heats at different rates because of the thermal radiation emitted by the two cans. Kit contains a lamp assembly with reflector and stand (bulb not included), one smooth metal can, one black metal can, and thermometer.
Specifications
- Description:Radiation can kit