Rectangular Refraction Cell
Rectangular Refraction Cell
470304-030
: RCRC01
High Quality for the Classroom.
- Clear acrylic
- Uniform wall thickness
This clear acrylic rectangular refraction cell is excellent for showing refraction and internal reflection in water, and to measure its refractive index. The cell has uniform wall thickness to eliminate distortions.
- Description:Rectangular Refraction Cell