Order Entry
Mexico
Orders LinkContactUsLinkComponent
Plastic Labware Value Set
Plastic Labware Value Set
Plastic Labware Value Set
Catalog # 470304-026
Supplier #:  PLSKIT2
Plastic Labware Value Set
Catalog # 470304-026
Plastic Labware Value Set
Plastic Labware Value Set
Plastic Labware Value Set
Price
$2,412.91
Log in for availability
Each (1UN)
Product Details & Documents
Economical Choice for the Classroom.
  • All items in the set with the exception of wash bottles are autoclavable
Economical, and convenient starter kit or refresher set. Includes an assortment of popular plastic labware items.
Ordering information: 5 Beakers (50, 100, 250, 500 and 1000 ml), 7 Graduated Cylinders (10, 25, 50, 100, 250, 500 and 1000 ml), 4 Wash Bottles (Unitary, 500 ml), 6 Storage Bottles (Square, Wide Mouth, 500 ml), 3 Erlenmeyer Flasks (Wide Mouth, with cap, 250 ml), 1 Test Tube Rack (with 12 plastic tubes)
Specifications
  • Description:
    Plastic Labware Value Set