Order Entry
Mexico
Orders LinkContactUsLinkComponent
Lab Tray
Lab Tray
Lab Tray
Catalog # 470303-962
Supplier #:  LTRAY1
Lab Tray
Catalog # 470303-962
Lab Tray
Lab Tray
Lab Tray
Price
$2,334.56
Log in for availability
Paquete de 10
Product Details & Documents
Ideal for containing small spills.
  • Low design takes up less space but still contains spills
  • Autoclavable
Flat polypropylene laboratory trays serve as a drip pan. Economical trays measuring 15×11.5" are versatile for general laboratory use.
Specifications
  • Description:
    Lab Tray