Detergent, LancerClean, LCD-P, Lancer
LancerClean Detergent, LCD-P, 2×2.5 gal.
Supplier: GETINGE USA SALES, LLC. MS
Supplier #: LCD2.5P
Detergent, LancerClean, LCD-P, Lancer
Catalog # MSPP-LCD25P
Supplier: GETINGE USA SALES, LLC. MS
Marketsource
Jump to:
Product Details & Documents
LCD-P is a potassium hydroxide based solution.
Provides excellent cleaning results for all types of contaminants and standard laboratory soils and also it can be used in all glassware washers.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:LancerClean Detergent, LCD-P
- Size:2×2.5 gal.