Order Entry
Mexico
ContactUsLinkComponent
 
Chart Paper, Thermo Scientific
Catalog # 55702-630
Supplier:  VWR International
Product available on GSA Advantage®
undefined
Chart Paper, Thermo Scientific
Catalog # 55702-630
Supplier:  VWR International
Product available on GSA Advantage®

Specifications

  • Description:
    Recorder chart paper, –40...+25 °C, 150 mm
  • Cat. no.:
    55702-630

Specifications

About this item

Circular chart paper, non-thermal.

  • Pressure-sensitive
  • 7 days

Range: −40...+25 °C