Carson HookUpz Universal Smartphone Binocular Adapter
Catalog # 470235-860
Supplier: CARSON OPTICAL
Specifications
- Description:Hookupz Smartphone Bino Adapter IB-700
- Cat. no.:470235-860
Specifications
About this item
The HookUpz Universal Smartphone Binocular Adapter from Carson easily connects your smartphone to most full sized binoculars.
- Objective Lens Diameter: 32 to 50 mm
Use your built-in camera application on your smartphone to take pictures and record video through the lenses of your binoculars.
Fits all phones. For specific information on your model, visit: www.carson.com/multicam.