Respirator Inlet Assemblies, 4100, MSA
10024-288
- Color:Black
- Description:Advantage® 4100 Facepiece Respirator Inlet Assembly; P100 Filtration Grade
- For use with:Advantage® 4100 Full-Facepiece Respirator
- Resistance:Asbestos
- Cat. no.:10024-288
Single-port facepiece is compatible with several MSA respirator models for comfortable and cost-efficient performance.
- Precision crafted to cover a wide variety of respiratory applications
- Standard nosecup reduces fogging
- Components lock into place in seconds for quick and convenient performance
Compatible with air-purifying respirators, supplied-air respirators, and powered air-purifying respirators. Features a harness and a standard nosecup for reduced fogging in low-temperature or high-moisture environments. Components lock into place in seconds for quick, convenient performance.