Order Entry
Mexico
Orders LinkContactUsLinkComponent
Solar Science Station
Solar Science Station
Solar Science Station
Catalog # 470234-660
Supplier:   BROWN DOG GADGETS
Número de proveedor #:  K1009
Solar Science Station
Catalog # 470234-660
Supplier:  BROWN DOG GADGETS
Solar Science Station
Solar Science Station
Solar Science Station
Price
$1,599.03
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Use solar power to teach STEAM with ease…while charging your USB device!
  • The kit can be assembled with only a screw driver and wire stripper, which means it is 100% reusable
The Solar Science Station is a non-soldering solar USB charger. Use it as a tool for recording solar data in the classroom and as a power supply for gadgets. We've designed it around a large 6V 200mA solar cell that sits on a rotating axis (with degree measures), so you can adjust the angle. We have also engraved degree measures on the side. Record heaps of scientific data by having students measure the angle in degrees, the time of day, and the voltage output on the LED Voltmeter display!
Ordering information: Parts include 6V 200 mA Solar Panel; Terminal Block; LED Voltmeter; USB Circuit; Banana Sockets; Laser Cut Wood Enclosure with Screws; and Optional Battery Pack.
Especificaciones
  • Description:
    Solar Science Station