Order Entry
Mexico
ContactUsLinkComponent
Cutting Board, High Density Polyethylene, Dynalon
Cutting Board, High Density Polyethylene, Dynalon
  490012-208
 :  DYNALON
undefined
Cutting Board, High Density Polyethylene, Dynalon
  490012-208
 :  DYNALON
 :  116405
  • Material:
    HDPE
  • Environmentally Preferable:
  • Cat. no.:
    490012-208
  • L×W×H:
    30.48 x 45.72 x 1.27 cm
  • Supplier No.:
    116405
Excellent surface for examining specimens, cutting, dissecting, etc.
This white cutting board series is made in the USA with an easy-to-clean surface. Provides excellent surface for examining specimens, cutting, dissecting, etc. NSF, FDA, and USDA approved material.